Merck
  • Low-viscosity matrix suspension culture enables scalable analysis of patient-derived organoids and tumoroids from the large intestine.
Low-viscosity matrix suspension culture enables scalable analysis of patient-derived organoids and tumoroids from the large intestine.
Communications biology (2021-09-15)
Yumiko Hirokawa, Jordan Clarke, Michelle Palmieri, Tao Tan, Dmitri Mouradov, Shan Li, Cong Lin, Fuqiang Li, Huijuan Luo, Kui Wu, Maree Faux, Chin Wee Tan, Margaret Lee, Grace Gard, Peter Gibbs, Antony W Burgess, Oliver M Sieber
ABSTRACT
Cell embedment into a solid support matrix is considered essential for the culture of intestinal epithelial organoids and tumoroids, but this technique presents challenges that impede scalable culture expansion, experimental manipulation, high-throughput screening and diagnostic applications. We have developed a low-viscosity matrix (LVM) suspension culture method that enables efficient establishment and propagation of organoids and tumoroids from the human large intestine. Organoids and tumoroids cultured in LVM suspension recapitulate the morphological development observed in solid matrices, with tumoroids reflecting the histological features and genetic heterogeneity of primary colorectal cancers. We demonstrate the utility of LVM suspension culture for organoid and tumoroid bioreactor applications and biobanking, as well as tumoroid high-throughput drug sensitivity testing. These methods provide opportunities for the study and use of patient-derived organoids and tumoroids from the large intestine.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
GenElute PCR Clean-Up Kit, sufficient for 70 purifications
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
TransPlex® Whole Transcriptome Amplification Kit, DNA polymerase separate.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture
Sigma-Aldrich
Kanamycin solution from Streptomyces kanamyceticus, 50 - 60 mg/mL in 0.9% NaCl, BioReagent, liquid, 0.1 μm filtered, suitable for cell culture