Merck
  • Antiviral treatment causes a unique mutational signature in cancers of transplantation recipients.
Antiviral treatment causes a unique mutational signature in cancers of transplantation recipients.
Cell stem cell (2021-09-09)
Jurrian K de Kanter, Flavia Peci, Eline Bertrums, Axel Rosendahl Huber, Anaïs van Leeuwen, Markus J van Roosmalen, Freek Manders, Mark Verheul, Rurika Oka, Arianne M Brandsma, Marc Bierings, Mirjam Belderbos, Ruben van Boxtel
ABSTRACT
Genetic instability is a major concern for successful application of stem cells in regenerative medicine. However, the mutational consequences of the most applied stem cell therapy in humans, hematopoietic stem cell transplantation (HSCT), remain unknown. Here we characterized the mutation burden of hematopoietic stem and progenitor cells (HSPCs) of human HSCT recipients and their donors using whole-genome sequencing. We demonstrate that the majority of transplanted HSPCs did not display altered mutation accumulation. However, in some HSCT recipients, we identified multiple HSPCs with an increased mutation burden after transplantation. This increase could be attributed to a unique mutational signature caused by the antiviral drug ganciclovir. Using a machine learning approach, we detected this signature in cancer genomes of individuals who received HSCT or solid organ transplantation earlier in life. Antiviral treatment with nucleoside analogs can cause enhanced mutagenicity in transplant recipients, which may ultimately contribute to therapy-related carcinogenesis.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, FITC conjugate, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Foscarnet sodium, British Pharmacopoeia (BP) Reference Standard
Sigma-Aldrich
Ganciclovir ready made solution, ≥99% (HPLC), 10 mM in DMSO
Sigma-Aldrich
Normal Mouse IgG, FITC conjugate, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunofluorescence.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%