Merck
  • Stress-induced nuclear condensation of NELF drives transcriptional downregulation.
Stress-induced nuclear condensation of NELF drives transcriptional downregulation.
Molecular cell (2021-02-07)
Prashant Rawat, Marc Boehning, Barbara Hummel, Fernando Aprile-Garcia, Anwit S Pandit, Nathalie Eisenhardt, Ashkan Khavaran, Einari Niskanen, Seychelle M Vos, Jorma J Palvimo, Andrea Pichler, Patrick Cramer, Ritwick Sawarkar
ABSTRACT
In response to stress, human cells coordinately downregulate transcription and translation of housekeeping genes. To downregulate transcription, the negative elongation factor (NELF) is recruited to gene promoters impairing RNA polymerase II elongation. Here we report that NELF rapidly forms nuclear condensates upon stress in human cells. Condensate formation requires NELF dephosphorylation and SUMOylation induced by stress. The intrinsically disordered region (IDR) in NELFA is necessary for nuclear NELF condensation and can be functionally replaced by the IDR of FUS or EWSR1 protein. We find that biomolecular condensation facilitates enhanced recruitment of NELF to promoters upon stress to drive transcriptional downregulation. Importantly, NELF condensation is required for cellular viability under stressful conditions. We propose that stress-induced NELF condensates reported here are nuclear counterparts of cytosolic stress granules. These two stress-inducible condensates may drive the coordinated downregulation of transcription and translation, likely forming a critical node of the stress survival strategy.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-RPB3, from rabbit, purified by affinity chromatography
Sigma-Aldrich
1,6-Hexanediol, 99%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets