Merck
  • Structural Determinants for Small-Molecule Activation of Skeletal Muscle AMPK α2β2γ1 by the Glucose Importagog SC4.
Structural Determinants for Small-Molecule Activation of Skeletal Muscle AMPK α2β2γ1 by the Glucose Importagog SC4.
Cell chemical biology (2018-04-17)
Kevin R W Ngoei, Christopher G Langendorf, Naomi X Y Ling, Ashfaqul Hoque, Swapna Varghese, Michelle A Camerino, Scott R Walker, Ylva E Bozikis, Toby A Dite, Ashley J Ovens, William J Smiles, Roxane Jacobs, He Huang, Michael W Parker, John W Scott, Mark H Rider, Richard C Foitzik, Bruce E Kemp, Jonathan B Baell, Jonathan S Oakhill
ABSTRACT
The AMP-activated protein kinase (AMPK) αβγ heterotrimer regulates cellular energy homeostasis with tissue-specific isoform distribution. Small-molecule activation of skeletal muscle α2β2 AMPK complexes may prove a valuable treatment strategy for type 2 diabetes and insulin resistance. Herein, we report the small-molecule SC4 is a potent, direct AMPK activator that preferentially activates α2 complexes and stimulates skeletal muscle glucose uptake. In parallel with the term secretagog, we propose "importagog" to define a substance that induces or augments cellular uptake of another substance. Three-dimensional structures of the glucose importagog SC4 bound to activated α2β2γ1 and α2β1γ1 complexes reveal binding determinants, in particular a key interaction between the SC4 imidazopyridine 4'-nitrogen and β2-Asp111, which provide a design paradigm for β2-AMPK therapeutics. The α2β2γ1/SC4 structure reveals an interaction between a β2 N-terminal α helix and the α2 autoinhibitory domain. Our results provide a structure-function guide to accelerate development of potent, but importantly tissue-specific, β2-AMPK therapeutics.
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Supelco
Phenformin hydrochloride, analytical standard
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline