Merck
  • SARS-CoV-2 infection rewires host cell metabolism and is potentially susceptible to mTORC1 inhibition.
SARS-CoV-2 infection rewires host cell metabolism and is potentially susceptible to mTORC1 inhibition.
Nature communications (2021-03-27)
Peter J Mullen, Gustavo Garcia, Arunima Purkayastha, Nedas Matulionis, Ernst W Schmid, Milica Momcilovic, Chandani Sen, Justin Langerman, Arunachalam Ramaiah, David B Shackelford, Robert Damoiseaux, Samuel W French, Kathrin Plath, Brigitte N Gomperts, Vaithilingaraja Arumugaswami, Heather R Christofk
ABSTRACT
Viruses hijack host cell metabolism to acquire the building blocks required for replication. Understanding how SARS-CoV-2 alters host cell metabolism may lead to potential treatments for COVID-19. Here we profile metabolic changes conferred by SARS-CoV-2 infection in kidney epithelial cells and lung air-liquid interface (ALI) cultures, and show that SARS-CoV-2 infection increases glucose carbon entry into the TCA cycle via increased pyruvate carboxylase expression. SARS-CoV-2 also reduces oxidative glutamine metabolism while maintaining reductive carboxylation. Consistent with these changes, SARS-CoV-2 infection increases the activity of mTORC1 in cell lines and lung ALI cultures. Lastly, we show evidence of mTORC1 activation in COVID-19 patient lung tissue, and that mTORC1 inhibitors reduce viral replication in kidney epithelial cells and lung ALI cultures. Our results suggest that targeting mTORC1 may be a feasible treatment strategy for COVID-19 patients, although further studies are required to determine the mechanism of inhibition and potential efficacy in patients.
Sigma-Aldrich
Torin2, ≥98% (HPLC)
Sigma-Aldrich
Everolimus, ≥95% (HPLC)
Sigma-Aldrich
Temsirolimus, ≥98% (HPLC)