- Hypertension in Relation to Dioxins and Polychlorinated Biphenyls from the Anniston Community Health Survey Follow-Up.
Hypertension in Relation to Dioxins and Polychlorinated Biphenyls from the Anniston Community Health Survey Follow-Up.
In 2014, we conducted a longitudinal study [Anniston Community Health Survey (ACHS II)] 8 y after the baseline (ACHS I). We investigated the relationship between persistent chlorinated compounds and hypertension in residents living around the former polychlorinated biphenyl (PCB) production plant in Anniston, Alabama. We also examined the potential role of inflammatory cytokines in those with hypertension. A total of 338 participants had their blood pressure measured and medications recorded, gave a blood sample, and completed a questionnaire. Prevalent hypertension was defined as taking antihypertensive medication or having systolic blood pressure >140 mmHg and/or diastolic pressure >90 mmHg; incident hypertension used similar criteria in those who developed hypertension since the baseline in 2005-2007. PCB congeners were categorized into structure-activity groups, and toxic equivalencies (TEQs) were calculated for dioxin-like compounds. Descriptive statistics, logistic and linear regressions, as well as Cox proportional hazard models, were used to analyze the associations between exposures and hypertension. Prevalent hypertension (78%) in ACHS II showed statistically significant adjusted odds ratios (ORs) for PCBs 74, 99, 138, 153, 167, 177, 183, and 187, ranging from 2.18 [95% confidence interval (CI): 1.10, 4.33] to 2.76 (95% CI: 1.14, 6.73), as well as for two estrogenic-like PCB groups, and the thyroid-like group [ORs ranging from 2.25 (95% CI: 1.07, 4.75) to 2.54 (95% CI: 1.13, 5.74)]. Furthermore, analysis of quartiles demonstrated a monotonic relationship for dioxin-like non-ortho (non-o)-PCB TEQs [fourth vs. first quartile: 3.66 (95% CI: 1.40, 9.56)]. Longitudinal analyses of incident hypertension supported those positive associations. The results were strongest for the di-o-PCBs [hazard ratio (HR)=1.93 (95% CI: 0.93, 4.00)] and estrogenic II PCB group [HR=1.90 (95% CI: 0.96, 3.78)] but were weaker for the dioxin TEQs. Findings supportive of positive associations were reported for dioxin-like mono-o- and non-o-PCBs as well as for nondioxin-like estrogenic and thyroid-like congeners with prevalent and incident hypertension, suggesting that multiple pathways may be involved in hypertension development. https://doi.org/10.1289/EHP5272.