Merck
  • N-acetylglucosamine drives myelination by triggering oligodendrocyte precursor cell differentiation.
N-acetylglucosamine drives myelination by triggering oligodendrocyte precursor cell differentiation.
The Journal of biological chemistry (2021-01-18)
Michael Sy, Alexander U Brandt, Sung-Uk Lee, Barbara L Newton, Judy Pawling, Autreen Golzar, Anas M A Rahman, Zhaoxia Yu, Graham Cooper, Michael Scheel, Friedemann Paul, James W Dennis, Michael Demetriou
ABSTRACT
Myelination plays an important role in cognitive development and in demyelinating diseases like multiple sclerosis (MS), where failure of remyelination promotes permanent neuro-axonal damage. Modification of cell surface receptors with branched N-glycans coordinates cell growth and differentiation by controlling glycoprotein clustering, signaling, and endocytosis. GlcNAc is a rate-limiting metabolite for N-glycan branching. Here we report that GlcNAc and N-glycan branching trigger oligodendrogenesis from precursor cells by inhibiting platelet-derived growth factor receptor-α cell endocytosis. Supplying oral GlcNAc to lactating mice drives primary myelination in newborn pups via secretion in breast milk, whereas genetically blocking N-glycan branching markedly inhibits primary myelination. In adult mice with toxin (cuprizone)-induced demyelination, oral GlcNAc prevents neuro-axonal damage by driving myelin repair. In MS patients, endogenous serum GlcNAc levels inversely correlated with imaging measures of demyelination and microstructural damage. Our data identify N-glycan branching and GlcNAc as critical regulators of primary myelination and myelin repair and suggest that oral GlcNAc may be neuroprotective in demyelinating diseases like MS.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC Antibody, Alexa Fluor 647 conjugate, clone mGalC, from mouse, ALEXA FLUOR 647
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®