Merck
Monodisperse drops templated by 3D-structured microparticles.
Science advances (2020-11-06)
Chueh-Yu Wu, Mengxing Ouyang, Bao Wang, Joseph de Rutte, Alexis Joo, Matthew Jacobs, Kyung Ha, Andrea L Bertozzi, Dino Di Carlo
ABSTRACT
The ability to create uniform subnanoliter compartments using microfluidic control has enabled new approaches for analysis of single cells and molecules. However, specialized instruments or expertise has been required, slowing the adoption of these cutting-edge applications. Here, we show that three dimensional-structured microparticles with sculpted surface chemistries template uniformly sized aqueous drops when simply mixed with two immiscible fluid phases. In contrast to traditional emulsions, particle-templated drops of a controlled volume occupy a minimum in the interfacial energy of the system, such that a stable monodisperse state results with simple and reproducible formation conditions. We describe techniques to manufacture microscale drop-carrier particles and show that emulsions created with these particles prevent molecular exchange, concentrating reactions within the drops, laying a foundation for sensitive compartmentalized molecular and cell-based assays with minimal instrumentation.
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%
Sigma-Aldrich
Poly(propylene glycol) diacrylate, average Mn ~800, contains 100 ppm BHT as inhibitor, 100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 575