Merck
  • H5N1 avian influenza virus induces apoptotic cell death in mammalian airway epithelial cells.
H5N1 avian influenza virus induces apoptotic cell death in mammalian airway epithelial cells.
Journal of virology (2008-09-13)
Tomo Daidoji, Takaaki Koma, Anariwa Du, Cheng-Song Yang, Mayo Ueda, Kazuyoshi Ikuta, Takaaki Nakaya
ABSTRACT
In recent years, the highly pathogenic avian influenza virus H5N1 has raised serious worldwide concern about an influenza pandemic; however, the biology of H5N1 pathogenesis is largely unknown. To elucidate the mechanism of H5N1 pathogenesis, we prepared primary airway epithelial cells from alveolar tissues from 1-year-old pigs and measured the growth kinetics of three avian H5 influenza viruses (A/Crow/Kyoto/53/2004 [H5N1], A/Duck/Hong Kong/342/78 [H5N2], and A/Duck/Hong Kong/820/80 [H5N3]), the resultant cytopathicity, and possible associated mechanisms. H5N1, but not the other H5 viruses, strongly induced cell death in porcine alveolar epithelial cells (pAEpC), although all three viruses induced similar degrees of cytopathicity in chicken embryonic fibroblasts. Intracellular viral growth and the production of progeny viruses were comparable in pAEpC infected with each H5 virus. In contrast, terminal deoxynucleotidyltransferase-mediated dUTP-biotin nick end labeling-positive cells were detected only in H5N1-infected pAEpC, and the activities of caspases 3, 8, and 9 were significantly elevated in pAEpC infected with H5N1, but not with H5N2 and H5N3. These results suggest that only H5N1 induces apoptosis in pAEpC. H5N1 cytopathicity was inhibited by adding the caspase inhibitor z-VAD-FMK; however, there were no significant differences in viral growth or release of progeny viruses. Further investigations using reverse genetics demonstrated that H5N1 hemagglutinin protein plays a critical role in inducing caspase-dependent apoptosis in infected pAEpC. H5N1-specific cytopathicity was also observed in human primary airway epithelial cells. Taken together, these data suggest that avian H5N1 influenza virus leads to substantial cell death in mammalian airway epithelial cells due to the induction of apoptosis.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%