Merck
Carbamylation of proteins in 2-D electrophoresis--myth or reality?
Journal of proteome research (2003-06-20)
John McCarthy, Femia Hopwood, David Oxley, Matthew Laver, Annalisa Castagna, Pier Giorgio Righetti, Keith Williams, Ben Herbert
ABSTRACT
Carbamylation is widely quoted as being a problem in 2-D gel analysis and the associated sample preparation steps. This modification occurs when iso-cyanate, a urea break-down product, covalently modifies lysine residues, thus inducing a change in isoelectric point. Urea is used at up to 9 M concentrations in sample preparation and 2-D gels because of its ability to disrupt protein structure and effect denaturation without the need for ionic surfactants such as SDS. We have studied carbamylation using 7 M urea and 2 M thiourea, under a range of experimental temperatures to establish when, and if, it occurs and what can be done to minimize the modification. The actual time required for protein extraction from a tissue is usually short compared to the time required for procedures such as reduction and alkylation and IPG rehydration and focusing. Therefore, it is the temperature during these post-extraction procedures that is the most critical factor. Our experiments have shown that carbamylation does not occur during electrophoresis in the presence of urea, even with prolonged run-times. However, under poorly controlled sample preparation and storage conditions, it can become a major event.
Sigma-Aldrich
Protein Extraction Reagent Type 4
Pricing and availability is not currently available.