Merck
  • Muscle ion transporters and antioxidative proteins have different adaptive potential in arm than in leg skeletal muscle with exercise training.
Muscle ion transporters and antioxidative proteins have different adaptive potential in arm than in leg skeletal muscle with exercise training.
Physiological reports (2017-10-19)
Magni Mohr, Tobias Schmidt Nielsen, Pál Weihe, Jákup A Thomsen, Giovanna Aquino, Peter Krustrup, Nikolai B Nordsborg
ABSTRACT
It was evaluated whether upper-body compared to lower-body musculature exhibits a different phenotype in relation to capacity for handling reactive oxygen species (ROS), H+, La-, Na+, K+ and also whether it differs in adaptive potential to exercise training. Eighty-three sedentary premenopausal women aged 45 ± 6 years (mean ± SD) were randomized into a high-intensity intermittent swimming group (HIS, n = 21), a moderate-intensity swimming group (MOS, n = 21), a soccer group (SOC, n = 21), or a control group (CON, n = 20). Intervention groups completed three weekly training sessions for 15 weeks, and pre- and postintervention biopsies were obtained from deltoideus and vastus lateralis muscle. Before training, monocarboxylate transporter 4 (MCT4), Na+/K+ pump α2, and superoxide dismutase 2 (SOD2) expressions were lower (P < 0.05) in m deltoideus than in m vastus lateralis, whereas deltoid had higher (P < 0.05) Na+/H+ exchanger 1 (NHE1) expression. As a result of training, Na+/K+ pump α2 isoform expression was elevated only in deltoideus muscle, while upregulation (P < 0.05) of the α1 and β1 subunits, phospholemman (FXYD1), NHE1, and superoxide dismutase 1 expression occurred exclusively in vastus lateralis muscle. The increased (P < 0.05) expression of MCT4 and SOD2 in deltoid muscle after HIS and vastus lateralis muscle after SOC were similar. In conclusion, arm musculature displays lower basal ROS, La-, K+ handling capability but higher Na+-dependent H+ extrusion capacity than leg musculature. Training-induced changes in the ion-transporting and antioxidant proteins clearly differed between muscle groups.
Sigma-Aldrich
Anti-Na+/H+ Exchanger-1 Antibody, CT, clone 4E9, clone 4E9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Monocarboxylate Transporter 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit