Merck
CoBATCH for High-Throughput Single-Cell Epigenomic Profiling.
Molecular cell (2019-09-01)
Qianhao Wang, Haiqing Xiong, Shanshan Ai, Xianhong Yu, Yaxi Liu, Jiejie Zhang, Aibin He
ABSTRACT
An efficient, generalizable method for genome-wide mapping of single-cell histone modifications or chromatin-binding proteins is lacking. Here, we develop CoBATCH, combinatorial barcoding and targeted chromatin release, for single-cell profiling of genomic distribution of chromatin-binding proteins in cell culture and tissue. Protein A in fusion to Tn5 transposase is enriched through specific antibodies to genomic regions, and Tn5 generates indexed chromatin fragments ready for library preparation and sequencing. Importantly, this strategy enables not only low-input epigenomic profiling in intact tissues but also measures scalable up to tens of thousands of single cells per experiment under both native and cross-linked conditions. CoBATCH produces ∼12,000 reads/cell with extremely low background. Mapping of endothelial cell lineages from ten embryonic mouse organs through CoBATCH allows for efficient deciphering of epigenetic heterogeneity of cell populations and cis-regulatory mechanisms. Thus, obviating specialized devices, CoBATCH is broadly applicable and easily deployable for single-cell profiling of protein-DNA interactions.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
TAPS, ≥99.5% (titration)
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)