Merck
Lipid droplets finally get a little R-E-S-P-E-C-T.
Cell (2009-12-01)
Robert V Farese, Tobias C Walther
ABSTRACT
Long underappreciated as important cellular organelles, lipid droplets are finally being recognized as dynamic structures with a complex and interesting biology. In light of this newfound respect, we discuss emerging views on lipid droplet biology and speculate on the major advances to come.
Product Description
Avanti
18:1-C11 TopFluor-C11 TopFluor TG, Avanti Research - A Croda Brand 810271P, powder
Avanti
18:1-18:1-C11 TopFluor TG, Avanti Research - A Croda Brand 810298C
Avanti
C4 TopFluor MG, Avanti Research - A Croda Brand 810293P, powder
Avanti
18:1-6:0 DNP-C11 TopFluor TG, Avanti Research - A Croda Brand 810272P, powder
Avanti
C11 TopFluor MG, Avanti Research - A Croda Brand 810296P, powder
Avanti
18:1-C11 TopFluor DG, Avanti Research - A Croda Brand 810297P, powder