Merck
A two-phase response of endothelial cells to hydrostatic pressure.
Journal of cell science (2018-06-01)
Valeria Prystopiuk, Benedikt Fels, Caroline Sophie Simon, Ivan Liashkovich, Dzmitry Pasrednik, Cornelius Kronlage, Roland Wedlich-Söldner, Hans Oberleithner, Johannes Fels
ABSTRACT
The vascular endothelium is exposed to three types of mechanical forces: blood flow-mediated shear stress, vessel diameter-dependent wall tension and hydrostatic pressure. Despite considerable variations of blood pressure during normal and pathological physiology, little is known about the acute molecular and cellular effects of hydrostatic pressure on endothelial cells. Here, we used a combination of quantitative fluorescence microscopy, atomic force microscopy and molecular perturbations to characterize the specific response of endothelial cells to application of pressure. We identified a two-phase response of endothelial cells with an initial response to acute (1 h) application of pressure (100 mmHg) followed by a different response to chronic (24 h) application. While both regimes induce cortical stiffening, the acute response is linked to Ca2+-mediated myosin activation, whereas the chronic cell response is dominated by increased cortical actin density and a loss in endothelial barrier function. GsMTx-4 and amiloride inhibit the acute pressure response, which suggests that the ENaC Na+ channel is a key player in endothelial pressure sensing. The described two-phase pressure response may participate in the differential effects of transient changes in blood pressure and hypertension.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Actinin antibody produced in mouse, clone BM-75.2, ascites fluid
Sigma-Aldrich
Anti-Filamin A Antibody, clone TI10, ascites fluid, clone TI10, Chemicon®
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-NUP62 antibody produced in mouse, clone 2D3, purified immunoglobulin, buffered aqueous solution