Merck
  • Novel approach to integrated DNA adductomics for the assessment of in vitro and in vivo environmental exposures.
Novel approach to integrated DNA adductomics for the assessment of in vitro and in vivo environmental exposures.
Archives of toxicology (2018-06-27)
Yuan-Jhe Chang, Marcus S Cooke, Chiung-Wen Hu, Mu-Rong Chao
ABSTRACT
Adductomics is expected to be useful in the characterization of the exposome, which is a new paradigm for studying the sum of environmental causes of diseases. DNA adductomics is emerging as a powerful method for detecting DNA adducts, but reliable assays for its widespread, routine use are currently lacking. We propose a novel integrated strategy for the establishment of a DNA adductomic approach, using liquid chromatography-triple quadrupole tandem mass spectrometry (LC-QqQ-MS/MS), operating in constant neutral loss scan mode, screening for both known and unknown DNA adducts in a single injection. The LC-QqQ-MS/MS was optimized using a representative sample of 23 modified 2'-deoxyribonucleosides reflecting a range of biologically relevant DNA lesions. Six internal standards (ISTDs) were evaluated for their ability to normalize, and hence correct, possible variation in peak intensities arising from matrix effects, and the quantities of DNA injected. The results revealed that, with appropriate ISTDs adjustment, any bias can be dramatically reduced from 370 to 8.4%. Identification of the informative DNA adducts was achieved by triggering fragmentation spectra of target ions. The LC-QqQ-MS/MS method was successfully applied to in vitro and in vivo studies to screen for DNA adducts formed following representative environmental exposures: methyl methanesulfonate (MMS) and five N-nitrosamines. Interestingly, five new DNA adducts, induced by MMS, were discovered using our adductomic approach-an added strength. The proposed integrated strategy provides a path forward for DNA adductomics to become a standard method to discover differences in DNA adduct fingerprints between populations exposed to genotoxins, and facilitate the field of exposomics.
Sigma-Aldrich
N2-Ethyl-2′-deoxyguanosine, ≥98% (HPLC), solid
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
2′-Deoxyuridine, ≥98.5%
Molsidomine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Ethynyl-2′-deoxycytidine, (EdC), AldrichCPR