
Zoledronic acid

Synonym(s):
Zoledronic acid monohydrate, P,P′-[1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid monohydrate, Zoledronate monohydrate, [1-hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl]bis(phosphonic acid) monohydrate
Empirical Formula (Hill Notation):
C5H10N2O7P2·H2O
CAS Number:
Molecular Weight:
290.10
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2