
Triflupromazine hydrochloride

Synonym(s):
N,N-Dimethyl-2-(trifluoromethyl)-10H-phenothiazine-10-propanamine monohydrochloride
Empirical Formula (Hill Notation):
C18H19F3N2S · HCl
CAS Number:
Molecular Weight:
388.88
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing