Skip to Content
Merck
Styrene-d8 ≥98 atom % D, ≥98% (CP), contains 4-t-butylcatechol as stabilizer

Styrene-d8

Styrene-d8 ≥98 atom % D, ≥98% (CP), contains 4-t-butylcatechol as stabilizer
Synonym(s):
Octadeuterostyrene
Linear Formula:
C6D5CD=CD2
CAS Number:
Molecular Weight:
112.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1