
Sodium cromoglicate for system suitability

Synonym(s):
Cromolyn sodium salt, Cromoglycate, Cromoglycic acid
Linear Formula:
C23H14O11Na2
CAS Number:
Molecular Weight:
512.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing