Skip to Content
Merck
Sodium 3-mercapto-1-propanesulfonate technical grade, 90%

Sodium 3-mercapto-1-propanesulfonate

Sodium 3-mercapto-1-propanesulfonate technical grade, 90%
Synonym(s):
3-Mercapto-1-propanesulfonic acid sodium salt, MPS
Linear Formula:
HSCH2CH2CH2SO3Na
CAS Number:
Molecular Weight:
178.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing