![(S)-(+)-1,2-Propanediol 96%](/deepweb/assets/sigmaaldrich/product/structures/319/830/07f6a9ac-834e-4163-b28a-d397f77ef355/640/07f6a9ac-834e-4163-b28a-d397f77ef355.png)
(S)-(+)-1,2-Propanediol
![(S)-(+)-1,2-Propanediol 96%](/deepweb/assets/sigmaaldrich/product/structures/319/830/07f6a9ac-834e-4163-b28a-d397f77ef355/640/07f6a9ac-834e-4163-b28a-d397f77ef355.png)
Synonym(s):
(S)-(+)-Propylene glycerol
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing