
Schisandrin

Synonym(s):
Schizandrin, (+)-Schizandrin, Schisandrin, Schisandrine, Wuweizi alcohol A, Wuweizichun A
Empirical Formula (Hill Notation):
C24H32O7
CAS Number:
Molecular Weight:
432.51
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing