
Schaftoside

Synonym(s):
8-α-L-Arabinopyranosyl-6-β-D-glucopyranosyl-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Apigenin 8-C-α-L-arabinoside 6-C-β-D-glucoside, Shaftoside
Empirical Formula (Hill Notation):
C26H28O14
CAS Number:
Molecular Weight:
564.49
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing