![Quinmerac PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/283/160/1883ce36-79c4-49f4-a1a4-2d35a499cdad/640/1883ce36-79c4-49f4-a1a4-2d35a499cdad.png)
Quinmerac
![Quinmerac PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/283/160/1883ce36-79c4-49f4-a1a4-2d35a499cdad/640/1883ce36-79c4-49f4-a1a4-2d35a499cdad.png)
Synonym(s):
7-Chloro-3-methyl-8-quinolinecarboxylic acid
Empirical Formula (Hill Notation):
C11H8ClNO2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2