
Pyrazosulfuron-ethyl

Synonym(s):
Ethyl 5-[(4,6-dimethoxy-2-pyrimidinyl)aminocarbonylaminosulfonyl]-1-methyl-1H-pyrazole-4-carboxylate
Empirical Formula (Hill Notation):
C14H18N6O7S
CAS Number:
Molecular Weight:
414.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2