
Poloxamer 407, micronized

Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:
Products
Product Category
Brand
pH Value
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing