Pinosylvin
Synonym(s):
(E)-3,5-Stilbenediol, (E)-5-(2-Phenylethenyl)-1,3-benzenediol, 5-Styrylresorcinol, trans-3,5-Dihydroxystilbene, Pinosylvine
Empirical Formula (Hill Notation):
C14H12O2
CAS Number:
Molecular Weight:
212.24
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing