
Phenethyl phenylacetate

Synonym(s):
Benzyl carbinyl phenylacetate
Linear Formula:
C6H5CH2CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
240.30
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing