
Palmitic acid-13C16

Synonym(s):
Hexadecanoic acid-13C16
Linear Formula:
13CH3(13CH2)1413CO2H
CAS Number:
Molecular Weight:
272.31
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing