
PAH Calibration Mix

Synonym(s):
Polynuclear Aromatic Hydrocarbons Mix
Products
Brand
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing