
Orphenadrine Related Compound C

Synonym(s):
Tofenacin hydrochloride, N-Methyl-2-[(2-methylphenyl)phenylmethoxy]ethanamine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2