
O-Acetyl-L-carnitine hydrochloride

Synonym(s):
(R)-3-Acetoxy-4-(trimethylammonio)butyrate hydrochloride, ALC, ALCAR, R-(−)-2-Acetyloxy-3-carboxy-N,N,N-trimethyl-1-propanaminium chloride
Empirical Formula (Hill Notation):
C9H17NO4 · HCl
CAS Number:
Molecular Weight:
239.70
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing