
N-Methoxy-N-methyl(triphenylphosphoranylidene)acetamide

Synonym(s):
(N-Methoxymethylaminocarbonylmethylene)triphenylphosphorane
Linear Formula:
(C6H5)3P=CHCON(OCH3)CH3
CAS Number:
Molecular Weight:
363.39
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing