
Metolazone for system suitability

Synonym(s):
Metolazone, 7-Chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo-6-quinazolinesulfonamide
Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing