
Metamizole Impurity B

Synonym(s):
4-Aminoantipyrine, 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone
Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1