Skip to Content
Merck
Lithium trimethylsilanolate 95%

Lithium trimethylsilanolate

Lithium trimethylsilanolate 95%
Synonym(s):
Trimethylsilanol lithium salt
Linear Formula:
(CH3)3SiOLi
CAS Number:
Molecular Weight:
96.13

Products

Product Category
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing