
Laurolactam

Synonym(s):
12-Aminododecanoic acid lactam, Aza-2-cyclotridecanone
Empirical Formula (Hill Notation):
C12H23NO
CAS Number:
Molecular Weight:
197.32
Products
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing