
L-Arginine ethyl ester dihydrochloride

Empirical Formula (Hill Notation):
C8H18N4O2 · 2HCl
CAS Number:
Molecular Weight:
275.18
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing