
Ketorolac Related Compound D

Synonym(s):
5-Benzoyl-2,3-dihydro-1H-pyrrolizine
Empirical Formula (Hill Notation):
C14H13NO
CAS Number:
Molecular Weight:
211.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2