
Glycopyrrolate Related Compound A

Synonym(s):
5-Nitroisophthalic acid, 5-Nitrobenzene-1,3-dicarboxylic acid
Linear Formula:
O2NC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
211.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing