Skip to Content
Merck
Fmoc-Phe(4-CF3)-OH ≥97% (HPLC)

Fmoc-Phe(4-CF3)-OH

Fmoc-Phe(4-CF3)-OH ≥97% (HPLC)
Synonym(s):
Fmoc-4-(trifluoromethyl)-L-phenylalanine
Empirical Formula (Hill Notation):
C25H20F3NO4
CAS Number:
Molecular Weight:
455.43

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing