
Flupyradifurone

Synonym(s):
4-[(6-Chloro-3-pyridylmethyl)(2,2-difluoroethyl)amino]furan-2(5H)-one
Empirical Formula (Hill Notation):
C12H11ClF2N2O2
CAS Number:
Molecular Weight:
288.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2