
Flunixin

Synonym(s):
2-[2-Methyl-3-(trifluoromethyl)phenylamino]nicotinic acid
Empirical Formula (Hill Notation):
C14H11F3N2O2
CAS Number:
Molecular Weight:
296.24
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing