
Eshmuno® CPX Resin

Products
Product Category
Brand
Physical Form
Product Line
Analyte Suitability
Separation Technique
Diameter (mm)
ID (mm)
Volume (ml)
Quality Segment
Application
Housing Material
Particle Size (µm)
Sterility
Showing 1-5 of 5
Compare
Product Number
Description
Pricing