Skip to Content
Merck
Epoxy embedding medium, hardener DDSA suitable for microscopy

Epoxy embedding medium, hardener DDSA

Epoxy embedding medium, hardener DDSA suitable for microscopy
Synonym(s):
Dodecenylsuccinic anhydride, DDSA
Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:

Products

Product Category
Formula Weight
Quality Segment
Application
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing