
EPA 610 Polynuclear Aromatic Hydrocarbons Mix

Synonym(s):
EPA 610 Polynuclear Aromatic Hydrocarbons Mixture
Products
Brand
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing