Skip to Content
Merck
DL-Glyceraldehyde 3-phosphate diethyl acetal barium salt

DL-Glyceraldehyde 3-phosphate diethyl acetal barium salt

DL-Glyceraldehyde 3-phosphate diethyl acetal barium salt
Synonym(s):
3,3-Diethoxy-1,2-propanediol 1-phosphate barium salt
Empirical Formula (Hill Notation):
C7H15BaO7P
CAS Number:
Molecular Weight:
379.49
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing