Skip to Content
Merck
(−)-Diisopropyl D-tartrate 98%

(−)-Diisopropyl D-tartrate

(−)-Diisopropyl D-tartrate 98%
Synonym(s):
D-(−)-Tartaric acid diisopropyl ester
Linear Formula:
[-CH(OH)CO2CH(CH3)2]2
CAS Number:
Molecular Weight:
234.25
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing