Skip to Content
Merck
Diethyl (difluoromethyl)phosphonate 97%

Diethyl (difluoromethyl)phosphonate

Diethyl (difluoromethyl)phosphonate 97%
Linear Formula:
F2CHP(O)(OC2H5)2
CAS Number:
Molecular Weight:
188.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing