
Dibenzyl N,N-diisopropylphosphoramidite

Linear Formula:
[(CH3)2CH]2NP(OCH2C6H5)2
CAS Number:
Molecular Weight:
345.42
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing